missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Histone Deacetylase 4/HDAC4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6205-100
This item is not returnable.
View return policy
Description
Histone Deacetylase 4/HDAC4 Monoclonal specifically detects Histone Deacetylase 4/HDAC4 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Histone Deacetylase 4/HDAC4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| HA6116, HD4EC 3.5.1.98, HDAC4, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
| E. coli-derived recombinant human Histone Deacetylase 4/HDAC4 Met1-Glu68 Accession # P56524 | |
| 100 μg | |
| Primary | |
| Detects human Histone Deacetylase 4/HDAC4 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 713802 | |
| Immunohistochemistry 5-25 ug/mL | |
| P56524 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9759 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction