missing translation for 'onlineSavingsMsg'
Learn More

Human Histone Deacetylase 4/HDAC4 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

196.00 € - 407.00 €

Specifications

Antigen Histone Deacetylase 4/HDAC4
Clone 713802
Dilution Immunohistochemistry 5-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752768
GREENER_CHOICE
View Documents
R&D Systems
MAB6205-SP
25 μg
196.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18716759
GREENER_CHOICE
View Documents
R&D Systems
MAB6205-100
100 μg
407.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone Deacetylase 4/HDAC4 Monoclonal specifically detects Histone Deacetylase 4/HDAC4 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Histone Deacetylase 4/HDAC4
Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
HA6116, HD4EC 3.5.1.98, HDAC4, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
E. coli-derived recombinant human Histone Deacetylase 4/HDAC4 Met1-Glu68 Accession # P56524
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Histone Deacetylase 4/HDAC4 in direct ELISAs.
713802
Immunohistochemistry
Unconjugated
Mouse
Human
P56524
9759
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.