missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB93454-SP
This item is not returnable.
View return policy
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Immunocytochemistry.
Specifications
| IFN-alpha | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
| HEK293 human embryonic kidney cell line transfected with human IFN-alpha 2, Met1-Glu188, Accession # P01563 | |
| 25 μg | |
| 3440 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Immunocytochemistry | |
| 995514 | |
| Immunocytochemistry 8-25 μg/mL | |
| P01563 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-alpha 2a,4a,6,7,8,10,16, and 17 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction