missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-alpha Antibody, R&D Systems™

Mouse Monoclonal Antibody
203.00 € - 466.00 €
Specifications
| Antigen | IFN-alpha |
|---|---|
| Clone | 995514 |
| Dilution | Immunocytochemistry 8-25 μg/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780557
![]() |
R&D Systems
MAB93454-SP |
25 μg |
203.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752217
![]() |
R&D Systems
MAB93454-100 |
100 μg |
466.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Human samples. It is validated for Immunocytochemistry.Specifications
| IFN-alpha | |
| Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P01563 | |
| 3440 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 995514 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
| HEK293 human embryonic kidney cell line transfected with human IFN-alpha 2, Met1-Glu188, Accession # P01563 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-alpha 2a,4a,6,7,8,10,16, and 17 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title