missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB9147-SP
This item is not returnable.
View return policy
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA, Immunohistochemistry.
Specifications
| IFN-epsilon | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020 | |
| E. coli-derived recombinant human IFN-epsilon Leu22-Arg208 Accession # Q86WN2 | |
| 25 μg | |
| 338376 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunohistochemistry | |
| 983338 | |
| Immunohistochemistry 1-25 ug/mL | |
| Q86WN2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human IFN-epsilon in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction