missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IFN-epsilon Antibody, R&D Systems™

Mouse Monoclonal Antibody
214.00 € - 408.00 €
Specifications
| Antigen | IFN-epsilon |
|---|---|
| Clone | 983338 |
| Dilution | Immunohistochemistry 1-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18717399
![]() |
R&D Systems
MAB9147-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744619
![]() |
R&D Systems
MAB9147-100 |
100 μg |
408.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
| IFN-epsilon | |
| Immunohistochemistry 1-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q86WN2 | |
| 338376 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 983338 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020 | |
| E. coli-derived recombinant human IFN-epsilon Leu22-Arg208 Accession # Q86WN2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IFN-epsilon in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title