Learn More
Human IFN-epsilon Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9147-100
 View more versions of this product
Product Code. 18744619
  • 408.00 € / 100µg
Description
IFN-epsilon Monoclonal specifically detects IFN-epsilon in Human samples. It is validated for ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IFN-epsilon
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020
E. coli-derived recombinant human IFN-epsilon Leu22-Arg208 Accession # Q86WN2
100 μg
338376
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Immunohistochemistry
983338
Immunohistochemistry 1-25 ug/mL
Q86WN2
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human IFN-epsilon in direct ELISAs.
Human
Supernatant