missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 18 publications
Brand: R&D Systems MAB200-SP
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-1 alpha/IL-1F1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P01583 | |
| IL1A | |
| E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
| 25 μg | |
| Primary | |
| Detects human IL-1 alpha/IL-1F1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-1 beta , -2, -3, -4, -5, -6, -7, -8, -9, -10, -11, -12, -13, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13. | |
| Human | |
| Purified |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| 4414 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3552 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur