missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 18 publications
226.00 € - 2155.00 €
Specifications
| Antigen | IL-1 alpha/IL-1F1 |
|---|---|
| Clone | 4414 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Neutralization, Immunocytochemistry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774317
![]() |
R&D Systems
MAB200-SP |
25 μg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18775759
![]() |
R&D Systems
MAB200-100 |
100 μg |
540.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789628
![]() |
R&D Systems
MAB200-500 |
500 μg |
2155.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).Specifications
| IL-1 alpha/IL-1F1 | |
| LYOPH | |
| Western Blot, Neutralization, Immunocytochemistry, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| IL1A | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 4414 | |
| Western Blot 1 ug/mL, Neutralization 0.05-0.15 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P01583 | |
| 3552 | |
| E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-1 alpha/IL-1F1 in ELISAs and Western blots. In ELISAs, this antibody does not cross-react with recombinant human (rh) IL-1 beta , -2, -3, -4, -5, -6, -7, -8, -9, -10, -11, -12, -13, rmIL-1 alpha , -1 beta , -3, -4, -5, -6, -7, -9, or -13. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title