missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB203-SP
This item is not returnable.
View return policy
Description
IL-3 Monoclonal specifically detects IL-3 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.
Specifications
| IL-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.03-0.08 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q6GS87 | |
| IL3 | |
| E. coli-derived recombinant human IL-3 Ala20-Phe152 Accession # Q6GS87 | |
| 25 μg | |
| Primary | |
| Detects human IL-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-3 is observed. Does not neutralize the biological activity of recombinant mouse IL-3. | |
| Human | |
| Purified |
| Western Blot, Neutralization, Immunocytochemistry | |
| 4806 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3562 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction