missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
214.00 € - 1695.00 €
Specifications
| Antigen | IL-3 |
|---|---|
| Clone | 4806 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.03-0.08 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Neutralization, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734319
![]() |
R&D Systems
MAB203-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18791938
![]() |
R&D Systems
MAB203-100 |
100 μg |
423.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799408
![]() |
R&D Systems
MAB203-500 |
500 μg |
1695.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-3 Monoclonal specifically detects IL-3 in Human samples. It is validated for Western Blot, Neutralization, Immunocytochemistry.Specifications
| IL-3 | |
| LYOPH | |
| Western Blot, Neutralization, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6GS87 | |
| 3562 | |
| E. coli-derived recombinant human IL-3 Ala20-Phe152 Accession # Q6GS87 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse IL-3 is observed. Does not neutralize the biological activity of recombinant mouse IL-3. |
| 4806 | |
| Western Blot 1 ug/mL, Neutralization 0.03-0.08 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Hematopoietic growth factor, IL3, IL-3MGC79398, interleukin 3 (colony-stimulating factor, multiple), interleukin-3, Mast cell growth factor, mast-cell growth factor, MCGF, MCGFMGC79399, MULTI-CSF, multilineage-colony-stimulating factor, Multipotential colony-stimulating factor, P-cell stimulating factor, P-cell-stimulating factor | |
| IL3 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title