missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Klotho Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5334-500
This item is not returnable.
View return policy
Description
Klotho Monoclonal specifically detects Klotho in Human samples. It is validated for Neutralization.
Specifications
| Klotho | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.2.1, EC 3.2.1.31, KL, klotho | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9365 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 775340 | |
| Neutralization 0.15-0.6 ug/mL | |
| Q9UEF7 | |
| KL | |
| Mouse myeloma cell line NS0-derived recombinant human Klotho Glu34-Ser981 Accession # Q9UEF7 | |
| 500 μg | |
| Primary | |
| Detects human Klotho in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Klotho (aa 35-982) is observed and no cross-reactivity with rmKlotho (aa 23-550), recombinant human (rh) Klotho beta (aa 53-997), or rhCytosolic beta-Glucosidase/GBA3 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction