missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Klotho Antibody, R&D Systems™

Rat Monoclonal Antibody
208.00 € - 2070.00 €
Specifications
| Antigen | Klotho |
|---|---|
| Clone | 775340 |
| Dilution | Neutralization 0.15-0.6 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741099
![]() |
R&D Systems
MAB5334-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777389
![]() |
R&D Systems
MAB5334-100 |
100 μg |
518.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752648
![]() |
R&D Systems
MAB5334-500 |
500 μg |
2070.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Klotho Monoclonal specifically detects Klotho in Human samples. It is validated for Neutralization.Specifications
| Klotho | |
| Neutralization 0.15-0.6 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.2.1, EC 3.2.1.31, KL, klotho | |
| KL | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 775340 | |
| Neutralization | |
| Unconjugated | |
| Rat | |
| Human | |
| Q9UEF7 | |
| 9365 | |
| Mouse myeloma cell line NS0-derived recombinant human Klotho Glu34-Ser981 Accession # Q9UEF7 | |
| Primary | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Detects human Klotho in direct ELISAs. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) Klotho (aa 35-982) is observed and no cross-reactivity with rmKlotho (aa 23-550), recombinant human (rh) Klotho beta (aa 53-997), or rhCytosolic beta-Glucosidase/GBA3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title