missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAMP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8731
This item is not returnable.
View return policy
Description
LAMP Monoclonal specifically detects LAMP in Human samples. It is validated for Neutralization.
Specifications
| LAMP | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ34254, FLJ35396, IgLON family member 3, IGLON3FLJ54658, LAMPFLJ37216, limbic system-associated membrane protein, LSAMP | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4045 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 110722 | |
| Neutralization 15 ug/mL | |
| AAC50569 | |
| LSAMP | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP Val29-Asn315 Accession # AAC50569 | |
| 500 μg | |
| Primary | |
| Detects human LAMP in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction