missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human LAMP Antibody, R&D Systems™

Mouse Monoclonal Antibody
195.00 € - 513.00 €
Specifications
| Antigen | LAMP |
|---|---|
| Clone | 110722 |
| Dilution | Neutralization 15 ug/mL |
| Applications | Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18795108
![]() |
R&D Systems
MAB8731-SP |
25 μg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159110
![]() |
R&D Systems
MAB8731 |
500 μg |
513.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LAMP Monoclonal specifically detects LAMP in Human samples. It is validated for Neutralization.Specifications
| LAMP | |
| Neutralization 15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ34254, FLJ35396, IgLON family member 3, IGLON3FLJ54658, LAMPFLJ37216, limbic system-associated membrane protein, LSAMP | |
| LSAMP | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 110722 | |
| Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAC50569 | |
| 4045 | |
| Mouse myeloma cell line NS0-derived recombinant human LAMP Val29-Asn315 Accession # AAC50569 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human LAMP in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title