missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melusin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2740
This item is not returnable.
View return policy
Description
Melusin/ITGB1BP2 Monoclonal specifically detects Melusin/ITGB1BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Melusin/ITGB1BP2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9UKP3 | |
| ITGB1BP2 | |
| E. coli-derived recombinant human Melusin Ser2-Glu347 Accession # Q9UKP3 | |
| 100 μg | |
| Primary | |
| Detects human Melusin in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 308705 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHORDC3, integrin beta 1 binding protein (melusin) 2, integrin beta-1-binding protein 2, ITGB1BP, ITGB1BP2, Melusin, MGC119214 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26548 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction