missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Melusin Antibody, R&D Systems™

Mouse Monoclonal Antibody
184.00 € - 378.00 €
Specifications
| Antigen | Melusin/ITGB1BP2 |
|---|---|
| Clone | 308705 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722837
![]() |
R&D Systems
MAB2740-SP |
25 μg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18143041
|
R&D Systems
MAB2740 |
100 μg |
378.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melusin/ITGB1BP2 Monoclonal specifically detects Melusin/ITGB1BP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Melusin/ITGB1BP2 | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9UKP3 | |
| 26548 | |
| E. coli-derived recombinant human Melusin Ser2-Glu347 Accession # Q9UKP3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Melusin in direct ELISAs and Western blots. |
| 308705 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHORDC3, integrin beta 1 binding protein (melusin) 2, integrin beta-1-binding protein 2, ITGB1BP, ITGB1BP2, Melusin, MGC119214 | |
| ITGB1BP2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title