missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Neuronal Pentraxin R/NPTXR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4414-100
This item is not returnable.
View return policy
Description
Neuronal Pentraxin R/NPTXR Monoclonal antibody specifically detects Neuronal Pentraxin R/NPTXR in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Neuronal Pentraxin R/NPTXR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| neuronal pentraxin receptor, NPR, NPTXR | |
| Mouse myeloma cell line, NS0-derived human Neuronal Pentraxin R/NPTXR, Ala24-Ala500, Accession # O95502 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 511527 | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| O95502 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23467 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction