missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Neuronal Pentraxin R/NPTXR Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 448.00 €
Specifications
| Antigen | Neuronal Pentraxin R/NPTXR |
|---|---|
| Clone | 511527 |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751277
![]() |
R&D Systems
MAB4414-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18773627
![]() |
R&D Systems
MAB4414-100 |
100 μg |
448.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neuronal Pentraxin R/NPTXR Monoclonal antibody specifically detects Neuronal Pentraxin R/NPTXR in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Neuronal Pentraxin R/NPTXR | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| neuronal pentraxin receptor, NPR, NPTXR | |
| Mouse myeloma cell line, NS0-derived human Neuronal Pentraxin R/NPTXR, Ala24-Ala500, Accession # O95502 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 511527 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O95502 | |
| 23467 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title