missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HMGB1/HMG-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB16901
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HMGB1/HMG-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3146 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 951420 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| P09429 | |
| HMGB1 | |
| Human HMGB1/HMG-1 synthetic peptide between amino acids Lys50-Ser100, Accession # P09429 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat HMGB1/HMG-1 in Western blot. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction