missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HMGB1/HMG-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00 € - 511.00 €
Specifications
| Antigen | HMGB1/HMG-1 |
|---|---|
| Clone | 951420 |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18761209
![]() |
R&D Systems
MAB16901-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752559
![]() |
R&D Systems
MAB16901 |
100 μg |
511.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| HMGB1/HMG-1 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| HMGB1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 951420 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Human, Mouse, Rat | |
| P09429 | |
| 3146 | |
| Human HMGB1/HMG-1 synthetic peptide between amino acids Lys50-Ser100, Accession # P09429 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human, mouse, and rat HMGB1/HMG-1 in Western blot. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title