missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myoglobin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB97204-100
This item is not returnable.
View return policy
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Myoglobin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MB, MGC13548, myoglobin, PVALB | |
| Purified human Myoglobin antigen from human heart, Accession No. P02144 | |
| 100 μg | |
| 4151.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| 2269D | |
| Western Blot 0.05 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL | |
| P02144 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Myoglobin in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction