missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Myoglobin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
170.00 € - 425.00 €
Specifications
| Antigen | Myoglobin |
|---|---|
| Clone | 2269D |
| Dilution | Western Blot 0.05 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723457
![]() |
R&D Systems
MAB97204-SP |
25 μg |
170.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754687
![]() |
R&D Systems
MAB97204-100 |
100 μg |
425.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myoglobin Monoclonal specifically detects Myoglobin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
| Myoglobin | |
| Western Blot 0.05 μg;/mL, ELISA, Immunohistochemistry 3-25 μg;/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P02144 | |
| 4151.0 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 2269D | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MB, MGC13548, myoglobin, PVALB | |
| Purified human Myoglobin antigen from human heart, Accession No. P02144 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Myoglobin in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title