Learn More
Human NKG2D/CD314 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 26 publications
Brand:  R&D Systems FAB139P-025
 View more versions of this product
Product Code. 18706407
  • 215.00 € / 25 tests
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NKG2D/CD314
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
KLRK1
BaF3 mouse pro-B cell line transfected with human NKG2D/CD314
25 Tests
Primary
Detects human NKG2D/CD314 in Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
149810
Flow Cytometry 10 uL/10^6 cells
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
22914
Human
Purified