missing translation for 'onlineSavingsMsg'
Learn More

Human NKG2D/CD314 PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 26 publications

215.00 € - 509.00 €

Specifications

Antigen NKG2D/CD314
Clone 149810
Dilution Flow Cytometry 10 uL/10^6 cells
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18706407
GREENER_CHOICE
View Documents
R&D Systems
FAB139P-025
25 Tests
215.00 €
25 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18749716
GREENER_CHOICE
View Documents
R&D Systems
FAB139P-100
100 Tests
509.00 €
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKG2D/CD314
Flow Cytometry 10 uL/10^6 cells
Monoclonal
Purified
RUO
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
22914
BaF3 mouse pro-B cell line transfected with human NKG2D/CD314
Primary
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
149810
Flow Cytometry
R-PE
Mouse
Human
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence
KLRK1
IgG1
Protein A or G purified from hybridoma culture supernatant
Detects human NKG2D/CD314 in Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.