Learn More
Human PD-1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems MAB11638-100
 View more versions of this product
Product Code. 30306035
  • 527.00 € / 100µg
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PD-1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Synthetic Peptide, Accession # Q15116
100 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Immunohistochemistry, Multiplex Immunofluorescence
2705E
Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL
Q15116
Rabbit
Protein A or G purified from cell culture supernatant
RUO
5133
Human
Purified