missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PD-1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
232.00 € - 527.00 €
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | 2705E |
| Dilution | Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL |
| Applications | Immunohistochemistry, Multiplex Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306010
|
R&D Systems
MAB11638-SP |
25 μg |
232.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306035
|
R&D Systems
MAB11638-100 |
100 μg |
527.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry,Multiplex ImmunofluorescenceSpecifications
| PD-1 | |
| Immunohistochemistry 1-10 μg/mL, Multiplex Immunofluorescence 15 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Synthetic Peptide, Accession # Q15116 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
| 2705E | |
| Immunohistochemistry, Multiplex Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q15116 | |
| 5133 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title