missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Peptide YY Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB9456-SP
This item is not returnable.
View return policy
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Immunohistochemistry.
Specifications
| Peptide YY | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Peptide tyrosine tyrosine, peptide YY, Peptide-YY, PYY, PYY1, PYY-I | |
| Human Peptide YY synthetic peptide Accession # P10082 | |
| 25 μg | |
| Primary | |
| Detects human Peptide YY. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 2163A | |
| Immunohistochemistry 3-10 ug/mL | |
| P10082 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 5697 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction