missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Peptide YY Antibody, R&D Systems™

Rabbit Monoclonal Antibody
195.00 € - 398.00 €
Specifications
| Antigen | Peptide YY |
|---|---|
| Clone | 2163A |
| Dilution | Immunohistochemistry 3-10 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18735408
![]() |
R&D Systems
MAB9456-SP |
25 μg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766738
![]() |
R&D Systems
MAB9456-100 |
100 μg |
398.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Immunohistochemistry.Specifications
| Peptide YY | |
| Immunohistochemistry 3-10 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Peptide tyrosine tyrosine, peptide YY, Peptide-YY, PYY, PYY1, PYY-I | |
| Human Peptide YY synthetic peptide Accession # P10082 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Peptide YY. |
| 2163A | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P10082 | |
| 5697 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title