missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-GSK-3 beta (S9) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
210.00 € - 407.00 €
Specifications
| Antigen | GSK-3 beta |
|---|---|
| Clone | 1253C |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736909
![]() |
R&D Systems
MAB8934-SP |
25 μg |
210.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702678
![]() |
R&D Systems
MAB8934 |
100 μg |
407.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSK-3 beta Monoclonal specifically detects GSK-3 beta in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
| GSK-3 beta | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunocytochemistry 1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 beta, glycogen synthase kinase-3 beta, GSK3 beta, GSK-3 beta, GSK3B, GSK3beta isoform | |
| GSK3B | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 1253C | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P49840 | |
| 2932 | |
| Phosphopeptide containing the human GSK-3 beta S9 site | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GSK-3 beta when phosphorylated at S9 in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title