missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Rab13 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8305
This item is not returnable.
View return policy
Description
Rab13 Monoclonal specifically detects Rab13 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Rab13 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P51153 | |
| RAB13 | |
| E. coli-derived recombinant human Rab13 Lys94-Thr191 Accession # P51153 | |
| 100 μg | |
| Primary | |
| Detects human Rab13 in direct ELISA and Western Blot. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 863028 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell growth-inhibiting gene 4 protein, growth-inhibiting gene 4 protein, RAB13, member RAS oncogene family, RAS-associated protein RAB13, ras-related protein Rab-13 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5872 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction