missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Rab13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
208.00 € - 405.00 €
Specifications
| Antigen | Rab13 |
|---|---|
| Clone | 863028 |
| Concentration | LYOPH |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18766959
![]() |
R&D Systems
MAB8305-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18159770
|
R&D Systems
MAB8305 |
100 μg |
405.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rab13 Monoclonal specifically detects Rab13 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Rab13 | |
| LYOPH | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P51153 | |
| 5872 | |
| E. coli-derived recombinant human Rab13 Lys94-Thr191 Accession # P51153 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Rab13 in direct ELISA and Western Blot. |
| 863028 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell growth-inhibiting gene 4 protein, growth-inhibiting gene 4 protein, RAB13, member RAS oncogene family, RAS-associated protein RAB13, ras-related protein Rab-13 | |
| RAB13 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title