missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8546-SP
This item is not returnable.
View return policy
Description
Reelin Monoclonal specifically detects Reelin in Human samples. It is validated for Immunohistochemistry.
Specifications
| Reelin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5649 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 940209 | |
| Immunohistochemistry 5-15 ug/mL | |
| P78509 | |
| RELN | |
| Chinese hamster ovary cell line CHO-derived recombinant human Reelin Ser1221-Gln2666 Accession # P78509 | |
| 25 μg | |
| Primary | |
| Detects human Reelin in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction