missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Reelin Antibody, R&D Systems™

Mouse Monoclonal Antibody
211.00 € - 413.00 €
Specifications
| Antigen | Reelin |
|---|---|
| Clone | 940209 |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742028
![]() |
R&D Systems
MAB8546-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18732218
![]() |
R&D Systems
MAB8546 |
100 μg |
413.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reelin Monoclonal specifically detects Reelin in Human samples. It is validated for Immunohistochemistry.Specifications
| Reelin | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.-, Reeler, reelin, RELN, RL, RLPRO1598 | |
| RELN | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 940209 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P78509 | |
| 5649 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Reelin Ser1221-Gln2666 Accession # P78509 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Reelin in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title