missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF9026
This item is not returnable.
View return policy
Description
Serpin A6/Cortisol Binding Globulin Polyclonal specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for Western Blot.
Specifications
| Serpin A6/Cortisol Binding Globulin | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| P08185 | |
| SERPINA6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Serpin A6 Met23-Val405 (Ser246Ala) Accession # P08185 | |
| 100 μg | |
| Primary | |
| Detects human Serpin A6 in direct ELISAs and Western Blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, and rhSerpin A4 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 6, CBG, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 866 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction