missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serpin A6 Antibody, R&D Systems™

Sheep Polyclonal Antibody
184.00 € - 459.00 €
Specifications
| Antigen | Serpin A6/Cortisol Binding Globulin |
|---|---|
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18707348
![]() |
R&D Systems
AF9026-SP |
25 μg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702629
![]() |
R&D Systems
AF9026 |
100 μg |
459.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serpin A6/Cortisol Binding Globulin Polyclonal specifically detects Serpin A6/Cortisol Binding Globulin in Human samples. It is validated for Western Blot.Specifications
| Serpin A6/Cortisol Binding Globulin | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| antitrypsin), member 6, CBG, CBGcorticosteroid-binding globulin, corticosteroid binding globulin, member 6, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, Serpin A6, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Transcortin | |
| SERPINA6 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot 1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| P08185 | |
| 866 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Serpin A6 Met23-Val405 (Ser246Ala) Accession # P08185 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human Serpin A6 in direct ELISAs and Western Blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, and rhSerpin A4 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title