missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A1/A2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB30192-SP
This item is not returnable.
View return policy
Description
Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Western Blot.
Specifications
| Serum Amyloid A1/A2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6288 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| 902728 | |
| Western Blot 0.2 ug/mL | |
| P0DJI8 | |
| Human SAA1 synthetic peptide Accession # P0DJI8 | |
| 25 μg | |
| Primary | |
| Detects human SAA1/SAA2 in direct ELISAs and Western blots. | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction