missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Amyloid A1/A2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 502.00 €
Specifications
| Antigen | Serum Amyloid A1/A2 |
|---|---|
| Clone | 902728 |
| Dilution | Western Blot 0.2 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18741379
![]() |
R&D Systems
MAB30192-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788349
![]() |
R&D Systems
MAB30192-100 |
100 μg |
502.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum Amyloid A1/A2 Monoclonal specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Western Blot.Specifications
| Serum Amyloid A1/A2 | |
| Western Blot 0.2 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P0DJI8 | |
| Human SAA1 synthetic peptide Accession # P0DJI8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SAA1/SAA2 in direct ELISAs and Western blots. |
| 902728 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 6288 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title