missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Smoothelin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8278
This item is not returnable.
View return policy
Description
Smoothelin Monoclonal specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| Smoothelin | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P53814 | |
| SMTN | |
| E. coli-derived recombinant human Smoothelin B Met1-Leu132 Accession # P53814 | |
| 100 μg | |
| Primary | |
| Detects human Smoothelin B in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| 885424 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ35365, FLJ38597, smoothelin, SMSMO, SMTN | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6525 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction