missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Smoothelin B Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 377.00 €
Specifications
| Antigen | Smoothelin |
|---|---|
| Clone | 885424 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Immunohistochemistry, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728379
![]() |
R&D Systems
MAB8278-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776949
![]() |
R&D Systems
MAB8278 |
100 μg |
377.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Smoothelin Monoclonal specifically detects Smoothelin in Human samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| Smoothelin | |
| LYOPH | |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| P53814 | |
| 6525 | |
| E. coli-derived recombinant human Smoothelin B Met1-Leu132 Accession # P53814 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Smoothelin B in direct ELISAs. |
| 885424 | |
| Immunohistochemistry 5-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ35365, FLJ38597, smoothelin, SMSMO, SMTN | |
| SMTN | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title