missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2766-SP
This item is not returnable.
View return policy
Description
SOX7 Monoclonal specifically detects SOX7 in Human samples. It is validated for Western Blot.
Specifications
| SOX7 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9BT81 | |
| SOX7 | |
| E. coli-derived recombinant human SOX7 Ser249-Ala345 Accession # Q9BT81 | |
| 25 μg | |
| Primary | |
| Detects human SOX7 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human SOX2, 3, 9, 10, or 17. | |
| Human | |
| Purified |
| Western Blot | |
| 288610 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83595 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction