missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
214.00 € - 405.00 €
Specifications
| Antigen | SOX7 |
|---|---|
| Clone | 288610 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18747489
![]() |
R&D Systems
MAB2766-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18103231
![]() |
R&D Systems
MAB2766 |
100 μg |
405.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX7 Monoclonal specifically detects SOX7 in Human samples. It is validated for Western Blot.Specifications
| SOX7 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BT81 | |
| 83595 | |
| E. coli-derived recombinant human SOX7 Ser249-Ala345 Accession # Q9BT81 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SOX7 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human SOX2, 3, 9, 10, or 17. |
| 288610 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7 | |
| SOX7 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title