missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SUMO3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2959R-500
This item is not returnable.
View return policy
Description
SUMO3 Monoclonal specifically detects SUMO3 in Human samples. It is validated for Immunocytochemistry.
Specifications
| SUMO3 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative | |
| SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6612 | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 401513R | |
| Immunocytochemistry 8-25 ug/mL | |
| P55854 | |
| SUMO3 | |
| E. coli-derived recombinant human SUMO3 Met1-Phe103 Accession No. P55854 | |
| 500 ug | |
| Primary | |
| Detects human SUMO3 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction