missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SUMO3 Antibody, R&D Systems™

Rat Monoclonal Antibody
224.00 € - 1780.00 €
Specifications
| Antigen | SUMO3 |
|---|---|
| Clone | 401513R |
| Dilution | Immunocytochemistry 8-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18743798
![]() |
R&D Systems
MAB2959R-SP |
25 μg |
224.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18713339
![]() |
R&D Systems
MAB2959R-100 |
100 μg |
448.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179610
|
R&D Systems
MAB2959R-500 |
500 ug |
1780.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18193011
|
R&D Systems
MAB2959R-01M |
1 mg |
1595.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SUMO3 Monoclonal specifically detects SUMO3 in Human samples. It is validated for Immunocytochemistry.Specifications
| SUMO3 | |
| Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| P55854 | |
| 6612 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Detects human SUMO3 in direct ELISAs. |
| 401513R | |
| Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| SMT3 (suppressor of mif two 3, yeast) homolog 1, SMT3 homolog 1, SMT3 suppressor of mif two 3 homolog 1, SMT3 suppressor of mif two 3 homolog 3 (S. cerevisiae), SMT3 suppressor of mif two 3 homolog 3 (yeast), SMT3A, SMT3B, SMT3H1, SMT3H1small ubiquitin-related modifier 3, SUMO-2, SUMO-3, ubiquitin-like protein SMT3A, Ubiquitin-like protein SMT3B | |
| SUMO3 | |
| Primary | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title