missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Synaptopodin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB8977-SP
This item is not returnable.
View return policy
Description
Synaptopodin Monoclonal specifically detects Synaptopodin in Human samples. It is validated for Immunohistochemistry.
Specifications
| Synaptopodin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1029, synaptopodin, SYNPO | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11346 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 918842 | |
| Immunohistochemistry 8-25 ug/mL | |
| Q8N3V7 | |
| SYNPO | |
| E. coli-derived recombinant human Synaptopodin Ser363-Gln490 Accession # Q8N3V7 | |
| 25 μg | |
| Primary | |
| Detects human Synaptopodin in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction