missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Synaptopodin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
208.00 € - 405.00 €
Specifications
| Antigen | Synaptopodin |
|---|---|
| Clone | 918842 |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18754408
![]() |
R&D Systems
MAB8977-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770397
![]() |
R&D Systems
MAB8977 |
100 μg |
405.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synaptopodin Monoclonal specifically detects Synaptopodin in Human samples. It is validated for Immunohistochemistry.Specifications
| Synaptopodin | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1029, synaptopodin, SYNPO | |
| SYNPO | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 918842 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N3V7 | |
| 11346 | |
| E. coli-derived recombinant human Synaptopodin Ser363-Gln490 Accession # Q8N3V7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Synaptopodin in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title