missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombopoietin/Tpo Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2881-SP
This item is not returnable.
View return policy
Description
Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot.
Specifications
| Thrombopoietin/Tpo | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 7066 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 34863 | |
| Western Blot 1 ug/mL | |
| P40225 | |
| THPO | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225 | |
| 25 μg | |
| Primary | |
| Detects human Thrombopoietin/Tpo in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Tpo is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction