missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Thrombopoietin/Tpo Antibody, R&D Systems™

Mouse Monoclonal Antibody
201.00 € - 542.00 €
Specifications
| Antigen | Thrombopoietin/Tpo |
|---|---|
| Clone | 34863 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751927
![]() |
R&D Systems
MAB2881-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18150001
![]() |
R&D Systems
MAB2881 |
500 μg |
542.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thrombopoietin/THPO Monoclonal specifically detects Thrombopoietin/THPO in Human samples. It is validated for Western Blot.Specifications
| Thrombopoietin/Tpo | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF | |
| THPO | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 34863 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P40225 | |
| 7066 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Thrombopoietin/Tpo Ser22-Gly353 Accession # P40225 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Thrombopoietin/Tpo in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse Tpo is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title