missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TL1A/TNFSF15 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB74421-100
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1 | |
| E. coli-derived recombinant human TL1A/TNFSF15 Leu72-Leu251 Accession # O95150 | |
| 100 μg | |
| Primary | |
| Detects human TL1A/TNFSF15 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Neutralization | |
| 2116D | |
| Western Blot 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Neutralization 0.04-0.2 ug/mL | |
| O95150 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 9966 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction