missing translation for 'onlineSavingsMsg'
Learn More

Human TL1A/TNFSF15 Antibody, R&D Systems™
GREENER_CHOICE

Rabbit Monoclonal Antibody

209.00 € - 494.00 €

Specifications

Antigen TL1A/TNFSF15
Clone 2116D
Dilution Western Blot 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Neutralization 0.04-0.2 ug/mL
Applications Western Blot, Immunohistochemistry, Neutralization
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18718799
GREENER_CHOICE
View Documents
R&D Systems
MAB74421-SP
25 μg
209.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18727079
GREENER_CHOICE
View Documents
R&D Systems
MAB74421-100
100 μg
494.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.
Specifications

Specifications

TL1A/TNFSF15
Western Blot 1 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Neutralization 0.04-0.2 ug/mL
Monoclonal
Rabbit
Human
O95150
9966
IgG
Protein A or G purified from cell culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
2116D
Western Blot, Immunohistochemistry, Neutralization
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1
E. coli-derived recombinant human TL1A/TNFSF15 Leu72-Leu251 Accession # O95150
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human TL1A/TNFSF15 in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.